A propos de cet article
Ergonomically designed, the Thermo Scientific™ Multifuge X4F/X4FR Pro series floor-standing centrifuges have a touch screen interface that enables easier programming for quicker results and ability to access and track all runs, rotor life and centrifuge health information for maximum versatility and power at every turn.
- ClickSeal™ bucket sealing system
- SMARTSpin™ technology
- Space-saving design
- Energy efficient
Secure locking system allows easy push-button installation and exchange of rotors. System saves time when switching between applications. Easy access to rotor chamber offers quick cleaning, a healthier working environment and longer unit life.
Biocontainment solution seals with a snap; eliminates screw caps and complicated clips. System has glove-friendly, one-handed open/close capability. System is certified by CAMR™ in Porton Down, UK.
Advanced rotor management system maximizes acceleration, braking and residual load imbalance for each rotor and bucket option. Technology optimizes safety and improves separations.
Floor standing option and compact dimensions optimize working area, the ergonomic height and curved design with motorized lid latch ensure easy sample loading and unloading, easy cleaning and promotes user safety.
System offers up to 40% energy consumption savings on industry standard protocols, such as standard blood separation or conical tubes processing.
Cell Culture package includes TX1000 rotor, set of four 1000 ml buckets, set of four ClickSeal™ biocontainment lids, set of four 10×50 ml conical tube adapters, set of four 24×15 ml conical tube adapters.
Blood Tube package includes TX1000 rotor, set of four 1000 ml buckets, set of four ClickSeal™ biocontainment lids, set of four 49×5/7 ml blood tube and 37×10 ml blood tube adapters.
Spécifications
- Catalog No:
- 76470-408
- 76470-434
- 76470-410
- 76470-432
- Model:
- Multifuge X4F (IVD certified)
- Multifuge X4F (IVD certified)
- Multifuge X4FR Pro (IVD certified)
- Multifuge X4FR Pro (IVD certified)
- Max. RCF - fixed angle rotor:
- 25830 ×g in Microliter 30×2
- 25830 ×g in Microliter 30×2
- 25830 ×g in Microliter 30×2
- 25830 ×g in Microliter 30×2
- Acceleration/braking profiles:
- Choice of 9 acceleration / 10 deceleration rates
- Choice of 9 acceleration / 10 deceleration rates
- Choice of 9 acceleration / 10 deceleration rates
- Choice of 9 acceleration / 10 deceleration rates
- Functions:
- SMARTSpin Imbalance Detection System; Auto-Lock III Rotor Locking System
- SMARTSpin Imbalance Detection System; Auto-Lock III Rotor Locking System
- SMARTSpin Imbalance Detection System; Auto-Lock III Rotor Locking System
- SMARTSpin Imbalance Detection System; Auto-Lock III Rotor Locking System
- Display:
- Full color touch screen interface
- Full color touch screen interface
- Full color touch screen interface
- Full color touch screen interface
- Voltage:
- 120 V
- 120 V
- 120 V
- 120 V
- W×D×H:
- 566×690×835 mm
- 566×690×835 mm
- 566×690×835 mm
- 566×690×835 mm