A propos de cet article
The Thermo Scientific™ Megafuge ST4F/ST4FR Plus floor-standing centrifuges offer intuitive controls and versatility for power at every turn.
- Auto-Lock rotor system
- ClickSeal™ bucket sealing system
- SMARTSpin™ technology
- Space-saving design
- Energy efficient
Secure locking system allows easy push-button installation and exchange of rotors. Biocontainment solution seals with a snap; eliminates screw caps and complicated clips. System has glove-friendly, one-handed open/close capability. System is certified by CAMR™ in Porton Down, UK.
Advanced rotor management system maximizes acceleration, braking and residual load imbalance for each rotor and bucket option. Technology optimizes safety and improves separations.
Floor standing option and compact dimensions optimize working area, the ergonomic height and curved design with motorized lid latch ensure easy sample loading and unloading, easy cleaning and promotes user safety.
Cell Culture package includes TX1000 rotor, set of four 1000 ml buckets, set of four ClickSeal™ biocontainment lids, set of four 10×50 ml conical tube adapters, set of four 24×15 ml conical tube adapters.
Blood Tube package includes TX1000 rotor, set of four 1000 ml buckets, set of four ClickSeal™ biocontainment lids, set of four 49×5/7 ml blood tube and 37×10 ml blood tube adapters.
Spécifications
- Catalog No:
- 76470-450
- CA76501-582
- CA76501-632
- CA76501-630
- CA76501-584
- 76470-436
- CA76501-624
- CA76501-622
- 76470-406
- 76470-404
- 76470-448
- 76470-446
- Model:
- Megafuge ST4F Plus
- Megafuge ST4F Plus
- Megafuge ST4F R Plus
- Megafuge ST4F Plus
- Megafuge ST4F R Plus
- Megafuge ST4F R Plus
- Megafuge ST4F R Plus
- Megafuge ST4F Plus
- Megafuge ST4F Plus
- Megafuge ST4F R Plus
- Megafuge ST4F Plus
- Megafuge ST4F R Plus
- Max. RCF - fixed angle rotor:
- 25830 ×g in Microliter 30×2
- 25830 ×g in Microliter 30×2
- 25830 ×g in Microliter 30×2
- 25830 ×g in Microliter 30×2
- 25830 ×g in Microliter 30×2
- 25830 ×g in Microliter 30×2
- 25830 ×g in Microliter 30×2
- 25830 ×g in Microliter 30×2
- 25830 ×g in Microliter 30×2
- 25830 ×g in Microliter 30×2
- 25830 ×g in Microliter 30×2
- 25830 ×g in Microliter 30×2
- Acceleration/braking profiles:
- Choice of 9 acceleration / 10 deceleration rates
- Choice of 9 acceleration / 10 deceleration rates
- Choice of 9 acceleration / 10 deceleration rates
- Choice of 9 acceleration / 10 deceleration rates
- Choice of 9 acceleration / 10 deceleration rates
- Choice of 9 acceleration / 10 deceleration rates
- Choice of 9 acceleration / 10 deceleration rates
- Choice of 9 acceleration / 10 deceleration rates
- Choice of 9 acceleration / 10 deceleration rates
- Choice of 9 acceleration / 10 deceleration rates
- Choice of 9 acceleration / 10 deceleration rates
- Choice of 9 acceleration / 10 deceleration rates
- Functions:
- SMARTSpin Imbalance Detection System; Auto-Lock III Rotor Locking System
- SMARTSpin Imbalance Detection System; Auto-Lock III Rotor Locking System
- SMARTSpin Imbalance Detection System; Auto-Lock III Rotor Locking System
- SMARTSpin Imbalance Detection System; Auto-Lock III Rotor Locking System
- SMARTSpin Imbalance Detection System; Auto-Lock III Rotor Locking System
- SMARTSpin Imbalance Detection System; Auto-Lock III Rotor Locking System
- SMARTSpin Imbalance Detection System; Auto-Lock III Rotor Locking System
- SMARTSpin Imbalance Detection System; Auto-Lock III Rotor Locking System
- SMARTSpin Imbalance Detection System; Auto-Lock III Rotor Locking System
- SMARTSpin Imbalance Detection System; Auto-Lock III Rotor Locking System
- SMARTSpin Imbalance Detection System; Auto-Lock III Rotor Locking System
- SMARTSpin Imbalance Detection System; Auto-Lock III Rotor Locking System
- Display:
- Push button and high contrast LCD interface
- Push button and high contrast LCD interface
- Push button and high contrast LCD interface
- Push button and high contrast LCD interface
- Push button and high contrast LCD interface
- Push button and high contrast LCD interface
- Push button and high contrast LCD interface
- Push button and high contrast LCD interface
- Push button and high contrast LCD interface
- Push button and high contrast LCD interface
- Push button and high contrast LCD interface
- Push button and high contrast LCD interface
- Voltage:
- 120 V
- 120 V
- 120 V
- 120 V
- 120 V
- 120 V
- 120 V
- 120 V
- 120 V
- 120 V
- 120 V
- 120 V
- W×D×H:
- 566×690×835 mm
- 566×690×835 mm
- 566×690×835 mm
- 566×690×835 mm
- 566×690×835 mm
- 566×690×835 mm
- 566×690×835 mm
- 566×690×835 mm
- 566×690×835 mm
- 566×690×835 mm
- 566×690×835 mm
- 566×690×835 mm