About this item
These borosilicate glass vials are a perfect fit for certain automated blood sampling systems. Available with pharmaceutical grade heparin, EDTA, citrate, or your choice of anticoagulant. Formulated for various blood volumes with a variety of additives. Can be customized.
- Maximize plasma or serum recovery from the smallest blood samples
- Anticoagulant type and amount are customizable
- Provided sterile
These extremely small (6×32 mm) borosilicate glass vials provide maximum plasma or serum recovery from the smallest blood samples, from 10 to 300 µl. Anticoagulant type and amount may be customized for your specific study needs.