DNA Model - Preassembled
# de catalogue 470224-018
Fournisseur: United Scientific Supplies
Spécifications
- Description:DNA model
- Cat. no.:470224-018
- Supplier No.:DNAM01
Spécifications
A propos de cet article
Built to scale.
- Easy to connect pieces
- Shows 16 base pairs
This pre-assembled DNA model is built to a scale of 10 cm to 1 nm and accurately depicts a 16 base pair section of DNA. The phosphatedeoxyribose backbone and the four bases are represented by custom plastic moldings that show the shapes of the molecules in scale and illustrate the bonding between them. The model is supported by a central pillar which can rotate on a circular stand.
Dimensions of the model are 24H×8" Ø, making it suitable for classroom demonstrations.
Caution: Small parts: Not for children under three years.
Customers Who Bought This Also Bought