Scalpel Handles, Integra
Supplier: Miltex
Certificates
About this item
EZ-OFF reusable plastic knife handles.
- Autoclavable
- Quick and easy removal of the blade
Fits mini blade (blades not available from Integra Miltex).
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.