Signs, 'NOTICE, FACE MASK REQUIRED EVEN FOR THOSE WITH COVID-19 VACCINATION', Accuform®
Fournisseur: ACCUFORM MANUFACTURING, INC
Certificats
A propos de cet article
Vaccination signs communicate health and safety policies, reinforce procedures, and identify locations.
- Multiple material and size options available
Choose from three material options. Adhesive Vinyl signs feature a printed matte surface with permanent acrylic adhesive back. These are moisture-resistant for a variety of light-duty indoor and sheltered outdoor marking needs. Plastic signs are durable with slight flex. Plastic is a good material choice for indoor and sheltered outdoor environments, as the printed surface has clear UV protective overcoat. Aluminum signs provide the strength of metal without rusting. They feature a printed surface protected by UV coating to withstand splashes, wash-downs, and outdoor conditions. They also offer abrasion and chemical resistance.
Customers Who Bought This Also Bought