A propos de cet article
SensiCare® Neoprene gloves offer a safe and economical alternative to natural rubber latex. Crafted from a second generation neoprene that exhibits outstanding performance characteristics.
- Smooth finish designed for tactile feel
- Brown color helps reduce glare from overhead lights in operating room
- Designed to enhance comfort
- Chemotherapy tested ASTM D6978-05
SensiCare® Neoprene surgical gloves provide industry-leading chemotherapy protection and a safe, economical alternative to natural rubber latex. Manufactured with an improved polycholorprene formulation, SensiCare® Neoprene produces a softer more elastic feel than traditional neoprene surgical gloves.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
- Catalog No:
- 76472-734
- 76472-722
- 76472-720
- 76472-730
- 76472-728
- 76472-726
- 76472-724
- 76472-732
- Latex/Latex-free:
- Latex-free
- Latex-free
- Latex-free
- Latex-free
- Latex-free
- Latex-free
- Latex-free
- Latex-free
- Powdered/Powder-free:
- Powder-free
- Powder-free
- Powder-free
- Powder-free
- Powder-free
- Powder-free
- Powder-free
- Powder-free
- Finish:
- Smooth
- Smooth
- Smooth
- Smooth
- Smooth
- Smooth
- Smooth
- Smooth
- Cuff style:
- Beaded
- Beaded
- Beaded
- Beaded
- Beaded
- Beaded
- Beaded
- Beaded
- Finger thickness:
- 7.8 mil
- 7.8 mil
- 7.8 mil
- 7.8 mil
- 7.8 mil
- 7.8 mil
- 7.8 mil
- 7.8 mil
- Sterility:
- Sterile
- Sterile
- Sterile
- Sterile
- Sterile
- Sterile
- Sterile
- Sterile