Epredia™ Peel-A-Way™ Disposable Embedding Molds, Richard-Allan Scientific
Fournisseur: Epredia
Certificats
A propos de cet article
Promote rapid paraffin embedding and hardening with Epredia™ Peel-A-Way disposable embedding molds.
Constructed of rigid transparent plastic.
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.