NEB Tube Opener, New England Biolabs
# de catalogue 76196-412
Fournisseur: New England Biolabs (NEB)
Spécifications
- Description:NEB tube opener, 2 each
- Cat. no.:76196-412
Spécifications
A propos de cet article
The NEB tube opener can be used to open a wide variety of microcentrifuge tubes.
Can be used for snap-on caps or screw-on caps.
Customers Who Bought This Also Bought