PCR Plates, Globe Scientific
Supplier: Globe Scientific
Certificates
About this item
Globe Scientific® PCR plates are offered in a variety of formats including no skirt, half skirt and full skirt variants in 0.1 ml (low profile) and 0.2 ml volumes. Available 384 well variants round out this expansive offering.
- Ultra-thin wall design (only 0.2 mm thick) for optimal temperture transfer
- Ultra-clear wells for easy sample visualization
- Easy-to-read alphanumeric indexing
- Manufactured from high-clarity, ultra-pure polypropylene (meets FDA 21 CFR 177.1520 & USP Class VI)
- Certified free of detectable DNase, RNase, Human DNA, PCR Inhibitors and tested Pyrogen-free
- Raised well rims for optimal sealing with strip caps, sealing mats or sealing film
Globe offers a PCR plate compatible most all commercially available PCR and qPCR instruments.
Customers Who Bought This Also Bought