ContactUsLinkComponent
Exel International Bulk, Unsterile Syringes, Air-Tite Products
Exel International Bulk, Unsterile Syringes, Air-Tite Products
  76436-742
 :  Air-Tite
Syringe, luer lock, bulk unsterile, quickly developing a reputation for providing quality products reasonable prices, large, bulk, unsterile syringe are convenient, low cost alternative when sterility is not issue, Manufactured in an ISO facility and packaged in a cleanroom environment, Size: 5ml
Exel International Bulk, Unsterile Syringes, Air-Tite Products
  76436-742
 :  Air-Tite
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
  • Capacity:
    5 ml
  • Description:
    Bulk unsterile syringes, Luer lock
  • Cat. no.:
    76436-742
The bulk, unsterile version of EXEL International's quality syringes. EXEL is known for unsurpassed quality disposable syringes. Great value at resonable costs.
  • Standard 3-part syringe
  • Latex- and pyrogen-free
  • Non-toxic
  • Positive plunger stop
  • Disposable
EXEL International is known for unsurpassed quality at a low price. The design includes bold print, clear barrels, and siliconized gaskets for smooth, consistent plunger motion and a positive plunger stop.
Certifications: Meets ISO standards.
Ordering information: Needles available separately.
Customers Who Bought This Also Bought