Y0105 Grain Goatskin Drivers with Cut Resistant Liner, Wells Lamont
Fournisseur: Wells Lamont
Certificats
A propos de cet article
Leather driver glove construction combines quality and comfort.
- Soft and flexible
- High level of cut, abrasion, and puncture protection
These key elements help reduce hand fatigue. Leather has natural abrasion resistant properties and a high level of tension strength. This increases the glove’s flexibility, creating greater control when handling and/or gripping objects.
Certifications: ANSI cut A5, ANSI abrasion 4, puncture level 3.
Ordering information: Sold by the dozen pairs.
Spécifications
- Catalog No:
- 76470-024
- 76470-026
- 76470-028
- 76470-030
- 76470-032
- 76470-034
- 76470-036
- Ambidextrous/Hand-specific:
- Hand-specific
- Hand-specific
- Hand-specific
- Hand-specific
- Hand-specific
- Hand-specific
- Hand-specific
- Disposable / reusable:
- Reusable
- Reusable
- Reusable
- Reusable
- Reusable
- Reusable
- Reusable
- Latex/Latex-free:
- Latex-free
- Latex-free
- Latex-free
- Latex-free
- Latex-free
- Latex-free
- Latex-free
- Powdered/Powder-free:
- Powder-free
- Powder-free
- Powder-free
- Powder-free
- Powder-free
- Powder-free
- Powder-free
- Liner material:
- HPPE/glass fibre
- HPPE/glass fibre
- HPPE/glass fibre
- HPPE/glass fibre
- HPPE/glass fibre
- HPPE/glass fibre
- HPPE/glass fibre
- Cuff style:
- Elastic
- Elastic
- Elastic
- Elastic
- Elastic
- Elastic
- Elastic