SP Bel-Art® Sterileware® Cross-Sectional Liquid Samplers, Bel-Art Products, a part of SP
Fournisseur: Bel-Art Products, a Part of SP
Certificats
A propos de cet article
The integrated valve of this sterile tool closes as it is pushed against a container bottom securing the sample for withdrawal. Sample can then be poured from the top of the tool into your sample container.
- Provides effective cross-sectional sampling of liquids and light creams
- Reaches bottom of container while keeping hand clear of container top
- Diameter allows for use through drum bunghole or other small openings
- Made from FDA and EU conforming materials; safe for food, drug and cosmetic use
- High density polyethylene (HDPE) samplers heat sealed in individual polyethylene bags
- Gamma irradiated sterile (SAL 10⁻⁶)
Lot stamped for accurate tracing; shipped with a Certificate of Processing for Sterilization.
Customers Who Bought This Also Bought