SP Bel-Art® Sterileware® Jumbo Sampling Pipettes, Bel-Art Products, a part of SP
Fournisseur: Bel-Art Products, a Part of SP
Certificats
A propos de cet article
Ideal for sampling from narrow- or wide-mouth containers up to 208-liter (55 Gallon) drums.
- Reaches into container while keeping hand clear of contents
- Diameter allows for use through drum bunghole or other small openings
- Color-coded end helps prevent handling errors
- Low-viscosity liquids enter easily through the 3 mm (¹/₈") tip hole
- Made from FDA and EU conforming materials; safe for food, drug and cosmetic use
- High density polyethylene (HDPE) pipettes heat sealed in individual polyethylene bags
- Gamma irradiated sterile (SAL 10⁻⁶)
Lot stamped for accurate tracing; shipped with a Certificate of Processing for Sterilization.
Customers Who Bought This Also Bought