SP Bel-Art Sterileware® Volumetric Sampling Spoons, Bel-Art Products, a part of SP
Fournisseur: Bel-Art Products, a Part of SP
Certificats
A propos de cet article
Suitable for use with powders, granules, liquids, gels or suspensions, these sterile spoons allow set volumes to be accurately measured and sampled.
- Leveling to top of bowl provides accurately measured samples
- Made from FDA and EU conforming materials; safe for food, drug and cosmetic use
- High density polyethylene (HDPE) spoons heat sealed in individual polyethylene bags
- Gamma irradiated sterile (SAL 10⁻⁶)
Lot stamped for accurate tracing; shipped with a Certificate of Processing for Sterilization.
Customers Who Bought This Also Bought