SP Bel-Art Sterileware® Cupped Powder Spatulas, Bel-Art Products, a part of SP
Supplier: Bel-Art Products, a Part of SP
Certificates
About this item
Available in both white and blue, these sterile spatulas are ideal for surface and general sampling of powders.
- Closed end keeps powders secure within spatula
- Chose contrasting color for easy visibility if left in material
- Made from FDA and EU conforming materials; safe for food, drug and cosmetic use
- White high impact polystyrene (HIPS), or blue polypropylene (PP) spatulas heat sealed in individual polyethylene bags
- Gamma irradiated sterile (SAL 10⁻⁶)
Lot stamped for accurate tracing; shipped with a certificate of processing for sterilization.
Frequently Bought Together