SP Bel-Art Sterileware® Triangular Scraper, Bel-Art Products, a part of SP
# de catalogue 76469-932
Fournisseur: Bel-Art Products, a Part of SP
Spécifications
- Length:22.5 cm (8⁷⁄₈")
- Cat. no.:76469-932
- Blade Width:110 mm (4⅜")
Spécifications
A propos de cet article
These sterile, plastic scrapers are perfect for cutting, cleaning and general scraping.
- Ideal for use in the manufacture and quality assurance of creams, ointments and pastes
- Made from FDA and EU conforming materials; safe for food, drug and cosmetic use
- High density polyethylene (HDPE) scrapers heat sealed in individual polyethylene bags
- Gamma irradiated sterile (SAL 10⁻⁶)
Lot stamped for accurate tracing; shipped with a Certificate of Processing for Sterilization.
Customers Who Bought This Also Bought