SP Bel-Art® Sterileware® Upright Handle Dippers or Ladles, Bel-Art Products, a part of SP
Fournisseur: Bel-Art Products, a Part of SP
Certificats
A propos de cet article
Available in blue or white to allow for effective sample contrast, these sterile ladles are useful for sampling liquids and creams.
- Upright handle allows for sampling from above container
- Curved handle end provides secure withdrawal of sample
- Fits into 2 in. openings such as a 45-gallon drum bunghole
- Made from FDA and EU conforming materials; safe for food, drug and cosmetic use
- White high density polyethylene (HDPE), or blue polypropylene (PP) ladles heat sealed in individual polyethylene bags
- Gamma irradiated sterile (SAL 10⁻⁶)
Lot stamped for accurate tracing; shipped with a Certificate of Processing for Sterilization.
Customers Who Bought This Also Bought