Spécifications
- Description:Ultimate plasticware set
- Cat. no.:470331-184
Spécifications
A propos de cet article
Durable and shatterproof 15 piece plastic labware set.
- Set includes 5 polypropylene beakers (50, 100, 250, 500, 1000 ml), 4 polypropylene graduated cylinders (10, 25, 50, 100 ml), 5 disposable pipettes (3 ml), 1 nylon bristle cleaning brush
- Beakers and graduated cylinders are made of high quality, chemically resistant polypropylene, feature excellent contact clarity, raised graduations, and a spout for drip free pouring
- Beakers and graduated cylinders are microwavable, autoclavable, and easy to clean with the included nylon bristle cleaning brush
- Disposable pipettes are graduated and made of LDPE
The Ultimate Plastics Kit features 15 pieces of essential plasticware. Beakers and graduated cylinders can be easily nested for compact storage. The set includes:
Five polypropylene beakers. Each beaker has raised graduations and features a spout for easy pouring, excellent contact clarity and very good chemical resistance.
• 50 ml (5 ml graduations) - 2.5" tall, 1.7" diameter
• 100 ml (5 ml graduations) - 2.75" tall, 2.1" diameter
• 250 ml (10 ml graduations) - 4" tall, 2.8" diameter
• 500 ml (10 ml graduations) - 4.75" tall, 3.6" diameter
• 1000 ml (20 ml graduations) - 5.8" tall, 4.4" diameter
Four polypropylene graduated cylinders. Each cylinder has excellent contact clarity and very good chemical resistance. Features include raised graduations, spouts for easy pouring and octagonal bases for extra stability.
• 10 ml (0.2 ml graduations) - 6" tall, 0.6" diameter, 1.6" base
• 25 ml (0.5 ml graduations) - 6.75" tall, 0.8" diameter, 2" base
• 50 ml (1 ml graduations) - 8" tall, 1" diameter, 2.3" base
• 100 ml (1 ml graduations) - 10" tall, 1.2" diameter, 2.7" base
Five LDPE, disposable pipettes. Pipettes have a 3 ml capacity and raised, 0.5 ml graduations. Each measures 6" in length and a 0.5" bulb diameter.
One nylon bristle cleaning brush that is perfect for scrubbing hard to reach corners of the included measuring cylinders and beakers. The brush has a twisted metal handle and measures 9" in total length. Bristles cover 3.75" of the wire handle and are 0.75" in diameter.