Fume Hood Worktops, Kewaunee
Fournisseur: Kewaunee
Certificats
A propos de cet article
Fume hood worktop, 304L stainless steel without cupsink.
Venturi bench hoods require a work top that must be ordered separately. Work tops are available in either kemresin or stainless steel, are dished ¹/₂" to retain spillage, and incorporate a 2" wide safety rim at the front. They may include cutouts for cupsinks or vent pipes when specified with the cutout option. Cupsinks must be ordered separately. Venturi floor founted hoods may be ordered with a removable work shelf and/or a work floor that must be ordered separately. Both are available in either kemresin or stainless steel, are dished ¹/₂" to retain spillage, incorporate a 2" wide safety rim at the front, and are not furnished with cutouts as standard.
Customers Who Bought This Also Bought