Glass Head with Duran® Flange and Three Necks, Ace Glass
Fournisseur: VWR International
Certificats
A propos de cet article
These Duran® flange glass heads are made with high-quality borosilicate glass.
- Duran® O-ring flange
- 24/40 center neck
- (2) 24/40 side necks
Featuring three in-line standard taper joints with a Duran® O-ring flange to ensure a tight seal with any of the matching 150 mm flanged vessels.
Customers Who Bought This Also Bought