VacTrap-G® (Glass) Vacuum Trap System, Foxx Life Sciences®
Fournisseur: Foxx Life Sciences
Certificats
A propos de cet article
Vactrap™ G (Glass) vacuum trap system, adher to the strict CDC recommendation for laboratory safety.
- Vactrap™ G includes the patented VersaCap® closure for tangle-free opening and closing
- Polypropylene VersaCap® has the highest temperature rating in the industry, 142 °C
- A 0.2 μm PTFE inline vent filter retains any aerosolized bacteria and protects the house vacuum or pump
- The entire assembly is nested neatly in a secondary container with a small footprint and incredible stability against tipping
- Pinch clamp allows vacuum adjustment
- Foxx EZFlow® TPE tubing with a high-level of shore hardness to eliminate kinking or collapsing under pressure
- Vactrap™ G uses next generation Foxx PUREGRIP® glass media bottles
- PUREGRIP® is made with BOROSIL® 3.3 Borosilicate Type 1 Glass which is mechanically strong, chemically resistant, and withstands thermal shock
The Foxx Life Sciences’ Vactrap™ G (Glass) vacuum trap system adheres to the strictest CDC recommendations for laboratory safety. With its new continuous full vacuum rating, it is ideal for protecting vacuum lines and users. Vactrap™ G safely contains waste produced during supernatant removal, chemical separation, and cell or tissue culture media aspiration.
Cap type: GL 45
Tubing size: ¼"
Vacuum rating: 1 bar
Certifications: Vactrap™ G is 40 CFR 264.175 compliant.
Customers Who Bought This Also Bought