A propos de cet article
Ultrapure molecular biology grade agarose pre-coated with GelGreen® Nucleic acid gel stain for convenient and safer preparation of precast gels.
- Non-toxic GelGreen® DNA dye is already in the agarose
- No need to handle concentrated dye solutions, for enhanced safety
- Use with TAE or TBE for 0.8 to 2% gels
- Low EEO, ultrapure molecular biology grade agarose
- Compatible with blue light illuminators
GelGreen® Agarose LE is our ultra-pure molecular biology grade LE agarose pre-coated with GelGreen® Nucleic acid gel stain. There is no need to handle concentrated fluorescent dye while preparing your gel, for greater convenience and safety. For agarose gels from 0.8 to 2% in TAE or TBE.
GelGreen® is a sensitive, stable and environmentally safe green fluorescent nucleic acid dye specifically designed for gel staining. GelGreen® is far more sensitive than SYBR® Safe. Unlike SYBR® dyes, which are known to be unstable, GelGreen® is very stable, both hydrolytically and thermally. GelGreen® has UV absorption between 250 and 300 nm and a strong absorption peak centered around 500 nm.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
- Catalog No:
- 76414-952
- 76414-954
- Environmentally Preferable:
- Safer Human and Environmental Health
- Safer Human and Environmental Health
- Environmental certification:
- Classified as non-hazardous to aquatic life under CCR title 22 regulations
- Classified as non-hazardous to aquatic life under CCR title 22 regulations