Standard Series Hard Hat, Bullard®
Catalogue # 76469-390
Fournisseur: Bullard
Spécifications
- Color:White
- Description:Standard series hard hat
- Cat. no.:76469-390
Spécifications
À propos de cet article
Featuring a four-point suspension, these hats are lightweight, low-profile, and have a low center of gravity, providing exceptional balance.
- Four-point suspension ratchet
- Cotton brow pad
- One size
General-purpose front-brim hard hats are non-vented and they have a front brim to protect the face from sun, reduce glare, and help shed rain. Standard series hard hats are made from durable polyethylene.
Certifications: Meets ANSI/ISEA Z89.1, Type I, Class E and G requirements.