Pliers, Needle Nose, Excelta
Fournisseur: Excelta
Certificats
A propos de cet article
The tapered jaws on these pliers allow better accessibility in tight places.
- Needle nose for tight areas
- Ergonomic handle, dual leaf springs
- Stainless steel construction-corrosion resistant
- Different sizes depending on application
Needle nose pliers have tapered noses for better accessibility in tight places. The noses aren't quite as strong as the 'Chain Nose' pliers due to the taper. Each of these is made of AISI 440 stainless steel to prevent oxidation. All include two leaf springs for more ergonomic control. They are offered in a small, medium or large head side, depending on the application. The 'D' specifies serrations on the tips for better control.
Customers Who Bought This Also Bought