Pliers, Bent Nose, Excelta
Supplier: Avantor
Certificates
About this item
Made of Stainless Steel, these bent nose pliers offer excellent visibility and accessibiity.
- Bent nose affords greater accessibility, visibility
- Ergonomic handle, dual leaf springs
- Stainless steel construction, corrosion resistant
- Different sizes depending on application
- Most available with serrated grips
Bent nose pliers are made of AISI 440 stainless steel and include two leaf springs for more ergonomic control. They are offered in a small, medium or large head side, depending on the application. Bent nose pliers offer greater accessability and more visibility when work in confined areas.
Customers Who Bought This Also Bought