Pliers, Chain Nose, Excelta
Fournisseur: Excelta
Certificats
A propos de cet article
These stainless steel general purpose pliers offer both smooth or serrated jaws.
- Ergonomic grips
- Stainless steel to retard oxidation
- Dual leaf springs
- Serrated tips available on most
Chain nose pliers are the most common style of pliers used. Each of these is made of AISI 440 stainless steel to prevent oxidation with the exception of the 744 which is made of carbon steel. All include two leaf springs for more ergonomic control. They are offered in a small, medium or large head side, depending on the application.
Customers Who Bought This Also Bought