HALO® PFAS Solutions, ENVIROCLASS, HPLC Columns, Advanced Materials Technology
Fournisseur: Advanced Materials Technology
Certificats
A propos de cet article
Designed for separation of novel and legacy short chain and long chain PFAS compounds containing branched and linear isomers, and with EPA methodology requirements in mind, HALO® PFAS offers a holistic solution.
- Application assured through method qualified lot analysis
- Optimal 2.7 µm Fused-Core® particle for rugged, reliable performance delivering high efficiency, low back pressure separations
- Endcapped alkyl phases for high sensitivity (no bleed) LCMS analysis
With both a PFAS-specific delay column optimized to prevent background PFAS contamination from interfering with the sample results and an analytical column for PFAS sample separation and detection, the HALO® PFAS solution delivers excellent selectivity, peak shape and necessary retention to perform fast, high-resolution separations in EPA methods 537.1, 533 and 8327.
The HALO® PFAS solution is different from other C18 offerings in that it is quality assured for PFAS analysis, providing confidence it will meet application demands.
Spécifications
- Catalog No:
- 76469-452
- 76469-454
- 76469-456
- 76469-458
- 76469-460
- 76469-462
- 76469-464
- 76469-466
- 76469-468
- 76469-470
- Endcapped:
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Max. pressure:
- 600 bar (9000 psi)
- 600 bar (9000 psi)
- 600 bar (9000 psi)
- 600 bar (9000 psi)
- 600 bar (9000 psi)
- 600 bar (9000 psi)
- 600 bar (9000 psi)
- 600 bar (9000 psi)
- 600 bar (9000 psi)
- 600 bar (9000 psi)
- pH range:
- 2 - 9
- 2 - 9
- 2 - 9
- 2 - 9
- 2 - 9
- 2 - 9
- 2 - 9
- 2 - 9
- 2 - 9
- 2 - 9
- Temperature limit:
- 60 °C (2 pH), 40 °C (9 pH)
- 60 °C (2 pH), 40 °C (9 pH)
- 60 °C (2 pH), 40 °C (9 pH)
- 60 °C (2 pH), 40 °C (9 pH)
- 60 °C (2 pH), 40 °C (9 pH)
- 60 °C (2 pH), 40 °C (9 pH)
- 60 °C (2 pH), 40 °C (9 pH)
- 60 °C (2 pH), 40 °C (9 pH)
- 60 °C (2 pH), 40 °C (9 pH)
- 60 °C (2 pH), 40 °C (9 pH)