Tweezers, Straight Fine Tips, Boley Style, Excelta Corp®
Fournisseur: Excelta
Certificats
A propos de cet article
Designed for general assembly.
- Strong tips for general assembly use
- Available in different materials for different applications
- Three different quality grades available
- Option of ergonomic grips
These 'BOLEY' straight medium point general purpose tweezer are offered in 3 different qualities. The higher the quality, the more precise the tips. They are offered in carbon steel (hard), stainless steel (rust-resistant) or stainless anti-magnetic steel (anti-magnetic). The 'ET' version features our Ergo-Tweeze® soft cushioned grips (10¹⁰ ohms/sq surface resistivity), which also protect the user from heat. The 'PH' suffix denotes plastic coated handles.
Customers Who Bought This Also Bought