A propos de cet article
When used with 95 kPa primaries, the BioCarrier acts as a leak-proof secondary and outer packaging in order to meet regulatory requirements. Intelsius® recommends the use of 95 kPa compliant pouches (available separately) if primaries are not 95 kPa.
- Rigid plastic case
- Multi-use
- Sample transport
- Class 6.2 Category B
- Temperature controlled
The Intelsius® BioCarrier range is a rigid, durable, reusable transit case designed for local courier routes and multiple site sample collections. The rugged polymer exterior with EPP insulated liner maintains sample integrity at 2 to 8 °C, and 15 to 25 °C for up to 24 hours.
Designed to address regular route, multiple site sample collection and 'home visit' temperature-sensitive injectable drug protection. The regulatory marked outer case features a clasp closure allowing the lid to be secured and the cases to be stacked and locked together with the insulated liner acting as a leak-proof secondary.
Available in two sizes, each system is regulatory compliant with pre-printed UN3373 labels for the shipment of Category B biological substances. Qualified for up to 24 hours, this product provides 2 to 8 °C thermal protection.
When using with 95 kPa primaries, this BioCarrier acts as a secondary and outer packaging in order to meet regulatory packing instructions. When primaries are not 95 kPa compliant, Intelsius® suggests the use of a 95 kPa secondary pouch.