Cutters, Standoff Shear, Large Frame, Excelta
Supplier: Excelta
Certificates
About this item
Reduces shock on delicate solder connections and sensitive components soldered into a printed circuit board.
- Anti-shock trimming of leads to specific length
- Ergonomic grips for comfort
- Double leaf springs for ease of use
This popular LARGE HEAD anti-shock shear cutter ensures that each and every lead is cut precisely to the desire length. There are many different configurations of this tool. Those with 'CUSTOM' suffixes require the buyer to specificy the desired lead length. They feature an ergonomically cushioned antimicrobial grip (10¹⁰ ohms/sq surface resistivity) with dual leaf springs. These tools are made of carbon tool steel and have flame hardened cutting edges.
Customers Who Bought This Also Bought