Brady® Inkjet Printable Repositionable Sign Sheets, Brady Worldwide
# de catalogue 76461-926
Fournisseur: Brady Worldwide
Spécifications
- Color:White
- Description:Sign sheets for ink jet printers, pack of 50
- Height:11"
- Material:Vinyl
- Cat. no.:76461-926
- Width:8.5"
Spécifications
A propos de cet article
Create the high-quality signage you need with your office inkjet printer with these Repositionable Sign sheets.
- Compatible surface types include doors, walls, windows
- Matte finish
- Repositionable acrylic adhesive type
The special aggressive adhesive on this Repositionable Inkjet Material stays in place when you need it to, but removes cleanly without leaving adhesive residue behind. Durable and flexible material making it ideal for areas where identification is frequently changed.
Customers Who Bought This Also Bought