Removable Polypropylene Labels, Brady
Fournisseur: Brady Worldwide
Certificats
A propos de cet article
These B-521 thermal transfer labels do not leave any adhesive residue behind when they are removed.
- Semi-gloss finish
- Removable acrylic adhesive type
Ideal for use in temporary labeling applications where solvent resistance and print performance are required. Have excellent low temperature resistance. If using this part in an IP series printer or i5100 printer, white print ribbon is not recommended.
Caution: Not recommended for long-term outdoor use.
Customers Who Bought This Also Bought