Starsys® 34" Tall Undercounter Storage Cart
Supplier: Metropolitan Wire
Certificates
About this item
Starsys® Undercounter Storage Carts enhance productivity and space efficiency.
- Durable easy-to- clean, polymer enclosures will not chip, peel, or rust
- Robust and stable structure accommodates even the most sensitive equipment
- Security options including non-locking, key locking, and electronic PIN locking with key override
The Starsys® Mobile Undercounter Cart is a polymer based, highly adaptive storage cart, that enhances productivity and space efficiency by empowering the end user with the ability to easily relocate, reposition, and transport stored contents as needed. This ability helps to create a more flexible workspace and to meet your changing workflow demands.
Microban® antimicrobial protection works 24/7 for an added level of protection against bacteria, mold and mildew that can cause stains, odors, and product degradation.
Customers Who Bought This Also Bought