SmartWall® Grid Shelf with Retaining Ledge
Fournisseur: Metropolitan Wire
Certificats
A propos de cet article
Create a unique storage and workstation system for often underutilized wall space.
- 2" high front retaining ledge helps to contain bulky items.
- 50 lbs. weight capacity
- Attaches quickly and easily to SmartWall® wire grids without using tools.
- Open-air wire design that promotes airflow.
Metroseal Gray epoxy finish keeps the shelves 'cleaner between cleanings' and protects the product from bacteria, mold, and fungi that cause stains, odor and product degradation.
Customers Who Bought This Also Bought