SmartWall® Light-Duty Shelf with Side Ledges
# de catalogue 76463-134
Fournisseur: Metropolitan Wire
Spécifications
- Finish:Metroseal epoxy
- Length:18½"
- Width:9½"
- Color:Gray
- Description:Light-duty shelf with side ledges
- Height:4⅝"
- Material:Steel
- Cat. no.:76463-134
Spécifications
A propos de cet article
Light-duty wire shelf with side ledges provides an efficient way to hold supplies.
- Ideal for managing small supplies or tools
- Easily attachable to SmartWall® grids without using tools
The Metroseal epoxy finish includes Microban® antimicrobials that keep shelves 'cleaner between cleaning'. This will work to prevent the growth of bacteria, mold and mildew.
Customers Who Bought This Also Bought