Super Erecta® Direct Wall Mount Single Shelf Bracket
Fournisseur: Metropolitan Wire
Certificats
A propos de cet article
Create a wall-mounted shelving system using Metro® Super Erecta® Shelving.
- Two supports are required per shelf
- Compatible with shelves with widths of 14", 18", and 24"
- Metroseal Gray epoxy finish for protection and visual appeal
These wall mounts are corrosion resistant and NSF approved for use in wet environments.
The epoxy coating works to keep the mounts cleaner for longer with the help of Microban® antimicrobial protection. Microban® prevents the growth of mold that can cause damages and odor.
Ordering information: Shelf sold separately.
Customers Who Bought This Also Bought