Kinesis® TELOS® neo™ MicroPlate™ SPE Populated 96-Well Microplates, Cole-Parmer®
Fournisseur: Cole-Parmer Instrument Co.
Certificats
A propos de cet article
Elution volume, 50 µl for minimal sample dilution and reduced evaporation time.
- Versatile 96-well format for small volume biological fluid samples
- Optimized well geometry for 5 and 10 mg sorbent masses
- Use with vacuum or positive pressure
- Polymer based bottom
The Int.Ø, frit design, and sorbent material of the TELOS® MicroPlate™ 96-well plates allow for maximum recovery with minimal sample dilution and reduced evaporation time. A well packed with 5 mg sorbent allows analytes to be eluted in as little as 50 µl.
Populated plate comes with 96 wells already in place. Modular design lets you simply remove any unwanted wells and add sealing strips. Full or partially populated plates can be processed using either vacuum or positive pressure.
Applications:
Mixed-mode strong anion exchange (PAX, PCX sorbent)
Mixed-mode weak anion exchange (WAX, WCX sorbent)
Non polar (PRP sorbent)
Customers Who Bought This Also Bought