Kinesis® TELOS® neo™ SPE Fixed 96-Well Microplates, Cole-Parmer®
Fournisseur: Cole-Parmer Instrument Co.
Certificats
A propos de cet article
Modular design lets you simply remove any unwanted wells and add sealing strips.
- Quickly obtain clean extracts and reduce time spent on method development
- Faster method development saves you money
- Polymer based bottom
- Water-wettable polymer is not affected by drying out
TELOS® neo™ populated plates come with 96 wells already in place ideal for high throughput assays. Polymer-based sorbents have no silanol interactions, excellent flow characteristics, and pH stability providing highly reproducible results. Elution volumes are kept to a minimum by allowing maximum interaction between the analyte and sorbent.
Applications:
Mixed-mode strong anion exchange (PAX, PCX sorbent)
Mixed-mode weak anion exchange (WAX, WCX sorbent)
Non polar (PRP sorbent)
Customers Who Bought This Also Bought