Specifications
- Glove Color:Natural
- Glove size:M
- Disposable / reusable:Disposable
- Latex/Latex-free:Latex-free
- Powdered/Powder-free:Powder-free
- Length:9.5"
- Sterility:Non sterile
- Cat. no.:76459-216
Specifications
About this item
This formulated blend of vinyl provides protection in low risk enviornments while maintaining protection, comfort, and dexterity.
- Soft fit and feel
- Latex-free and powder-free
- DOP and DEHP (phthalate) free
- Smooth inner coating for easy donning, and to help avoid irritation
- Applicable for many industries
Suitable alternative for individuals with Type I allergies. Great for a wide range of applications including medical, examination, healthcare, nursing, laboratory, IT, general cleaning, and industrial use.
Meets ASTM D5250 standards for poly-vinyl chloride gloves for medical applications. Meets EN 374 and EN 455 standards. FDA 510k certified and meets food handling requirements.