SmartWall® Tray Drying Racks, Metro™
Fournisseur: Metropolitan Wire
Certificats
A propos de cet article
SmartWall® Tray Drying Racks can easily create a fast, sanitary drying system.
- Ideal for trays, sheet pans, and large lids
- 50 lbs. weight capacity
Metroseal finish is suitable for wet and damp environments and includes Microban® antimicrobial product protection to inhibit the growth of bacteria, mold, mildew, and fungus that cause odors and product degradation.
Customers Who Bought This Also Bought