Super Erecta® Wall Mount Shelving Post with Brackets, Metro™
Fournisseur: Metropolitan Wire
Certificats
A propos de cet article
Combine with shelves and shelf supports to create a post-type wall mounted shelving solution.
- 13" posts are ideal for 1-shelf tier configurations
- 33" posts support 2-shelf configurations
- 54" posts can accommodate 3 or more shelving tiers
- 63" posts are designed to handle 4 or more shelving tiers
Corrosion resistant Metroseal Gray epoxy finish features Microban® antimicrobial production protection to keep the product 'cleaner between cleanings'.
Customers Who Bought This Also Bought