Super Erecta® Storage Basket for Wire Shelving and SmartWall® Wall Shelving, Metro™
Fournisseur: Metropolitan Wire
Certificats
A propos de cet article
Generous-sized baskets attach in seconds to hanger rails, wire shelves, or wall grids.
- Available in three different sizes
- Baskets are compatible with the SmartWall® grid system and Super Erecta® Shelving
- Hang in place easily with two hooks that attach to any wire shelving unit or grid without using tools
Metroseal options come with built-in Microban® antimicrobial additives. With this addition layer of protection, this product is safe from bacteria, mold, mildew and fungus which will increase its life span and keep it clean longer.
Customers Who Bought This Also Bought