Scissors, Hard Wire Carbide Insert Blades, Excelta Corp®
Fournisseur: Excelta
Certificats
A propos de cet article
These slim stainless steel scissors has tungsten carbide insert blades for cutting tough materials.
- Cuts small hard wires, (carbide insert)
- Ergonomic grips, dual leaf springs
- Available in different head sizes
Ideal for hard wire cutting applications.
Customers Who Bought This Also Bought