Dissecting Forceps
Fournisseur: DR INSTRUMENTS INC. SE
Certificats
A propos de cet article
Quality fine point forceps.
- Straight or bent tip
- Stainless steel construction
- Textured grip
Dissecting forceps feature straight fine points. Made with high quality stainless steel. Fine points are great for delicate dissection purposes and can be used under the microscope.
Caution: These instruments are intended for training and educational use only. These are not intended for human or animal procedures.
Customers Who Bought This Also Bought