SP Wilmad-LabGlass Solid State NMR Rotor Cap Removers, SP Industries
Fournisseur: SP Industries
Certificats
A propos de cet article
Easily remove end caps or base plugs.
- Zero-turn operation
- Manufactured from stainless steel
Eliminates possible damage to the cap and rotor.
Customers Who Bought This Also Bought