SP Wilmad-LabGlass Autosampler Low Pressure / Vacuum NMR Tubes, SP Industries
Fournisseur: SP Industries
Certificats
A propos de cet article
Manufactured to instrument specifications.
- Robust sealing system allows pressure build-up inside the sample
- Works with Bruker autosamplers
- Axial symmetric design guarantees its application in spinning experiments
These low pressure tubes are optimized for automatic samplers that require clearance on sample tube height. The length of this system after tightening the piston and removing the top glass vacuum adapter is 199 ±1 mm; the bottom NMR tube is 137 ±1 mm.
Customers Who Bought This Also Bought