SP Wilmad-LabGlass PTFE Syringe Needles for NMR Tube Filling, SP Industries
Supplier: SP Industries
Certificates
About this item
Provides easy sample loading for NMR tubes.
- Offered in various diameters to fit even the smallest tubes
- Fits all Luer lock syringes
Ideal for washing NMR tubes.
Frequently Bought Together