SP Wilmad-LabGlass Bruker X Band High Temperature EPR Tube, SP Industries
# de catalogue 76459-978
Fournisseur: SP Industries
Spécifications
- Length:203 mm (8")
- O.D.:4 mm
- Wall thickness:0.4 mm
- Description:200 psi
- Material:Suprasil
Spécifications
A propos de cet article
This EPR sample tube was made specifically for Bruker’s ER 4114HT-1000 cavity.
- The ⁷/₁₅ taper joint fits to Wilmad's DWGSK3352 high temperature dewar insert
Three wall finger system made from synthetic quartz yields low thermal expansion coefficient, which results in only a small resonance frequency drift due to wall heating.
Customers Who Bought This Also Bought