SP Wilmad-LabGlass Precision High Pressure Vacuum EPR Tubes, SP Industries
Fournisseur: SP Industries
Certificats
A propos de cet article
This high pressure/vacuum tube is a highly reliable EPR tube for pressure experiments.
- Valve is made of PTFE and opened by turning counterclockwise
Designed to connect to a ¹/₈" metal (SST or brass) vacuum line using Swagelok® fittings or a rubber vacuum hose.
Caution: Caution should be exercised when using at elevated or reduced pressures as performance of tubes under pressure or vacuum cannot be guaranteed. Adequate safety shielding should always be used when working in these conditions.
Customers Who Bought This Also Bought